Micro-burette with Schellbach stripe and glass key, class AS, DURAN®
Supplier: DWK Life Sciences
243201108 243201605 243202207
612-5219EA 160 EUR
612-5219 612-5220 612-5221
Micro-burette with Schellbach stripe and glass key, class AS, DURAN®
Burettes
DURAN®, borosilicate glass 3.3, clear.
  • With Schellbach stripe and main graduations as circular divisions
  • Straight stopcock
  • With glass key
  • Marks and inscription in blue
  • Calibration is based on the poured out volume ('Ex')
With printed retrace code (batch identification).
Certifications: Complies to the DIN ISO 385.