4-Hydroxybenzaldehyde, Sigma-Aldrich®
Supplier: Merck
Warning
54590-250G-F 54590-50G-F
SIAL54590-250G-FEA 127 EUR
SIAL54590-250G-F SIAL54590-50G-F
4-Hydroxybenzaldehyde, Sigma-Aldrich®
4-Hydroxybenzaldehyde
Formula: C₇H₆O₂
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 116 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 174 °C
Storage Temperature: Ambient
MDL Number: MFCD00006939
CAS Number: 123-08-0
EINECS: 204-599-1
Merck Index: 13,04836