4-Phenoxybenzaldehyde ≥98%
Supplier: Thermo Fisher Scientific
Synonyms:
p-Phenoxybenzenecarbaldehyde, 4-Phenoxybenzenecarbaldehyde
B22329.03
B22329.06
B22329.14
B22329.03EA
35
EUR
B22329.03
B22329.06
B22329.14
4-Phenoxybenzaldehyde ≥98%
4-Phenoxybenzaldehyde
Purity: 98%
CAS No.: 67-36-7
Molecular Formula: C13H10O2
Formula Weight: 198.22
Flash Point:
Formula:
C₁₃H₁₀O₂ Boiling Pt: 184…186 °C (14 mmHg) Melting Pt: 24…25 °C Density: 1.132 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003383 CAS Number: 67-36-7 EINECS: 200-650-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...