2,6-Dimethoxybenzonitrile

Supplier: Thermo Scientific

SB01246.1 SB01246.10 SB01246.50
MAYBSB01246.1EA 0 EUR
MAYBSB01246.1 MAYBSB01246.10 MAYBSB01246.50
2,6-Dimethoxybenzonitrile
2,6-Dimethoxybenzonitrile
Formula: C₉H₉NO₂
Boiling Pt: 310 °C (1013 hPa)
MDL Number: MFCD00001788
CAS Number: 16932-49-3
EINECS: 241-000-2
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR