Sodium benzoate, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Benzoic acid sodium salt
M-Clarity™ quality level = MQ200
71300-1KG
71300-250G
SIAL71300-1KGEA
54.7
EUR
SIAL71300-1KG
SIAL71300-250G
Sodium benzoate, Sigma-Aldrich®
Sodium benzoate
Formula:
C₆H₅COONa Melting Pt: 410…430 °C Density: 1.44 g/cm³ (20 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012463 CAS Number: 532-32-1 EINECS: 208-534-8 REACH: 01-2119460683-35 Merck Index: 13,08654 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...