3-Hydroxy-4-nitrobenzoic acid
Supplier: Thermo Scientific
|
Warning
|
SB01945.1
SB01945EA
SB01945.50
MAYBSB01945.1EA
0
EUR
MAYBSB01945.1
MAYBSB01945EA
MAYBSB01945.50
3-Hydroxy-4-nitrobenzoic acid
3-Hydroxy-4-nitrobenzoic acid
Formula:
C₇H₅NO₅ Melting Pt: 234…235 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007110 CAS Number: 619-14-7 EINECS: 210-580-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...