2,6-Dichlorobenzaldehyde ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
A13295.22
A13295.14
A13295.36
A13295.22EA
67
EUR
A13295.22
A13295.14
A13295.36
2,6-Dichlorobenzaldehyde ≥98%
2,6-Dichlorobenzaldehyde
Purity: 98%
CAS No.: 83-38-5
Molecular Formula: C7H4Cl2O
Formula Weight: 175.01
Flash Point: 135°(275°F)
Melting Point: 69-73°
Boiling Point: 124°/10mm
Beilstein Reference: 386477
UN No.: 3261
MDL No.: MFCD00003307
Formula:
C₇H₄Cl₂O MW: 175.01 g/mol Boiling Pt: 124 °C (10 mmHg) Melting Pt: 69…73 °C Flash Pt: 135 °C (275 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003307 CAS Number: 83-38-5 EINECS: 201-472-2 ADR: 8 III |