2-Chlorobenzaldehyde, Sigma-Aldrich®

Supplier: Merck
Danger

124974-100G 124974-500G
SIAL124974-100GEA 51.1 EUR
SIAL124974-100G SIAL124974-500G
2-Chlorobenzaldehyde, Sigma-Aldrich®
2-Chlorobenzaldehyde
Formula: C₇H₅ClO
MW: 140.57 g/mol
Boiling Pt: 211.9
Melting Pt: 10
Density: 1.248 g/cm³ (20 °C)
Flash Pt: 88
Storage Temperature: Ambient
MDL Number: MFCD00003304
CAS Number: 89-98-5
EINECS: 201-956-3
UN: 3265
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR