3-Chlorobenzaldehyde
Supplier: Thermo Scientific
|
Warning
|
SB01128.1
SB01128.10
SB01128.50
MAYBSB01128.1EA
0
EUR
MAYBSB01128.1
MAYBSB01128.10
MAYBSB01128.50
3-Chlorobenzaldehyde
3-Chlorobenzaldehyde
Formula:
C₇H₅ClO MW: 140.57 g/mol Boiling Pt: 214 Melting Pt: 17…18 °C Density: 1.241 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003350 CAS Number: 587-04-2 EINECS: 209-596-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...