4-Chlorobenzaldehyde ≥98%
Supplier: Thermo Fisher Scientific
Warning
A12757.30 A12757.0B A12757.0I
A12757.30EA 39.2 EUR
A12757.30 A12757.0B A12757.0I
4-Chlorobenzaldehyde ≥98%
4-Chlorobenzaldehyde
Purity: 98% CAS No.: 104-88-1 Molecular Formula: C7H5ClO Formula Weight: 140.57 Flash Point: 88°(190°F) Odour: Pungent Boiling Point: 213-214° Beilstein Reference: 385858 UN No.: 3077 MDL No.: MFCD00003379
Formula: C₇H₅ClO
MW: 140.57 g/mol
Boiling Pt: 213…214 °C
Melting Pt: 45…49 °C
Density: 1.196 g/cm³ (20 °C)
Flash Pt: 88 °C (190 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003379
CAS Number: 104-88-1
EINECS: 203-247-4
UN: 3077
ADR: 9 III