4-Chlorobenzonitrile
Supplier: Thermo Scientific
Warning
SB00343.1 SB00343.10 SB00343.50
MAYBSB00343.1EA 0 EUR
MAYBSB00343.1 MAYBSB00343.10 MAYBSB00343.50
4-Chlorobenzonitrile
4-Chlorobenzonitrile
Formula: C₇H₄ClN
Boiling Pt: 223 °C (1013 hPa)
Melting Pt: 90…93 °C
Density: 1,1133 g/cm³ (16.8 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001813
CAS Number: 623-03-0
EINECS: 210-765-4