2,3-Dichlorobenzonitrile ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
A14809.06
A14809.14
A14809.06EA
84
EUR
A14809.06
A14809.14
2,3-Dichlorobenzonitrile ≥98%
2,3-Dichlorobenzonitrile
Purity: 98%
CAS No.: 6574-97-6
Molecular Formula: C7H3Cl2N
Formula Weight: 172.01
Melting Point: 60-65°
Boiling Point: 114°/6mm
UN No.: 3439
MDL No.: MFCD00016372
Formula:
C₇H₃Cl₂N MW: 172.01 g/mol Boiling Pt: 114 °C (6 mmHg) Melting Pt: 60…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016372 CAS Number: 6574-97-6 UN: 3439 ADR: 6.1 III |