Dichlobenil
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2,6-Dichlorobenzonitrile
SB00156.1
SB00156.10
SB00156.50
MAYBSB00156.1EA
0
EUR
MAYBSB00156.1
MAYBSB00156.10
MAYBSB00156.50
Dichlobenil
Dichlobenil
Formula:
C₇H₃Cl₂N MW: 172.01 g/mol Boiling Pt: 279 Melting Pt: 145…146 °C Density: 1.623 g/cm³ (–101 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001781 CAS Number: 1194-65-6 EINECS: 214-787-5 UN: 3276 ADR: 6.1 III Merck Index: 13,03068 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...