3,4-Dichlorobenzonitrile
Supplier: Thermo Scientific
BTB06163.1
BTB06163.10
MAYBBTB06163.1EA
0
EUR
MAYBBTB06163.1
MAYBBTB06163.10
3,4-Dichlorobenzonitrile
3,4-Dichlorobenzonitrile
Formula:
C₇H₃Cl₂N MW: 172.01 g/mol Melting Pt: 86…89 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016379 CAS Number: 6574-99-8 EINECS: 229-494-8 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...