Blood lancets, BD Microtainer® contact-activated
Supplier: BD Medical
366594 366594UK 366592 366593
BDAM366594EA 129 EUR
BDAM366594 BDAM366594UK BDAM366592 BDAM366593
Blood lancets, BD Microtainer® contact-activated
Blood Lancets
BD Microtainer® contact-activated blood lancet is a safety-engineered device used for fingerstick collection of blood.
  • Enhanced user comfort
  • Activates on contact
  • Simplifies the procedure
Caution: Fingersticks should not be performed on children less than one year of age according to CLSI guidelines.