4-Chloro-1,3-difluorobenzene ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
A15495.09
A15495.18
A15495.30
A15495.09EA
34.2
EUR
A15495.09
A15495.18
A15495.30
4-Chloro-1,3-difluorobenzene ≥98%
4-Chloro-1,3-difluorobenzene
Purity: 98%
CAS No.: 1435-44-5
Molecular Formula: C6H3ClF2
Formula Weight: 148.54
Flash Point: 32°(89°F)
Refractive Index: 1.4750
Odour: Aromatic
Melting Point: -26°
Boiling Point: 126-127°
Beilstein Reference: 2081077
UN No.: 1993
MDL No.: MFCD00042569
Formula:
C₆H₃ClF₂ Boiling Pt: 126…127 °C Melting Pt: –26 °C Density: 1.353 Flash Pt: 32 °C (89 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042569 CAS Number: 1435-44-5 ADR: 3,III |