1-Chloro-4-fluorobenzene 98%
Supplier: Thermo Fisher Scientific
Warning
A13690.14 A13690.22 A13690.36
A13690.14EA 43.4 EUR
A13690.14 A13690.22 A13690.36
1-Chloro-4-fluorobenzene 98%
1-Chloro-4-fluorobenzene
Refractive Index: 1.4950
Beilstein Reference: 1904542
Formula: C₆H₄ClF
Boiling Pt: 129…130 °C
Melting Pt: –22 °C
Density: 1.228
Flash Pt: 29 °C (84 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000603
CAS Number: 352-33-0
EINECS: 206-521-1
ADR: 3,III