1-Chloro-4-iodobenzene ≥99%
Supplier: Thermo Fisher Scientific
|
Warning
|
A13459.14
A13459.22
A13459.36
A13459.14EA
45.3
EUR
A13459.14
A13459.22
A13459.36
1-Chloro-4-iodobenzene ≥99%
1-Chloro-4-iodobenzene
Purity: 99%
CAS No.: 637-87-6
Molecular Formula: C6H4ClI
Formula Weight: 238.46
Flash Point: 108°(226°F)
Melting Point: 52-56°
Boiling Point: 226-227°
Beilstein Reference: 1634414
MDL No.: MFCD00001053
Formula:
C₆H₄ClI MW: 238.45 g/mol Boiling Pt: 226…227 °C Melting Pt: 52…56 °C Density: 1.886 g/cm³ (57 °C) Flash Pt: 108 °C (226 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001053 CAS Number: 637-87-6 EINECS: 211-305-5 |