2,4-Dichloroaniline ≥99%
Supplier: Thermo Fisher Scientific
|
Danger
|
A10518.0E
A10518.22
A10518.30
A10518.36
A10518.0EEA
448
EUR
A10518.0E
A10518.22
A10518.30
A10518.36
2,4-Dichloroaniline ≥99%
2,4-Dichloroaniline
Purity: 99%
CAS No.: 554-00-7
Molecular Formula: C6H5Cl2N
Formula Weight: 162.02
Flash Point: 149°(300°F)
Melting Point: 59-63°
Boiling Point: 242°
Beilstein Reference: 386422
UN No.: 3442
MDL No.: MFCD00007661
Formula:
C₆H₅Cl₂N Boiling Pt: 242 °C Melting Pt: 59…63 °C Density: 1.567 Flash Pt: 149 °C (300 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007661 CAS Number: 554-00-7 EINECS: 209-057-8 UN: 3442 ADR: 6.1,II |