2,5-Dichloroaniline ≥99%
Supplier: Thermo Fisher Scientific
Danger
A11151.0E A11151.30 A11151.36
A11151.0EEA 277 EUR
A11151.0E A11151.30 A11151.36
2,5-Dichloroaniline ≥99%
2,5-Dichloroaniline
Purity: 99% CAS No.: 95-82-9 Molecular Formula: C6H5Cl2N Formula Weight: 162.02 Flash Point: 139°(282°F) Melting Point: 48-52° Boiling Point: 250-251° Beilstein Reference: 1447438 UN No.: 3442 MDL No.: MFCD00007667
Formula: C₆H₅Cl₂N
MW: 162.02 g/mol
Boiling Pt: 250…251 °C
Melting Pt: 48…52 °C
Density: 1.54
Flash Pt: 139 °C (282 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007667
CAS Number: 95-82-9
EINECS: 202-455-2
UN: 3442
ADR: 6.1 II