Bottles, wide neck, HDPE, round, sealable, with screw cap and sealing eyelets
Supplier: BURKLE
0322-1000 0322-0250 0322-0500 0322-2000
BURK0322-1000EA 6.8 EUR
BURK0322-1000 BURK0322-0250 BURK0322-0500 BURK0322-2000
Bottles, wide neck, HDPE, round, sealable, with screw cap and sealing eyelets
Bottles Plastic Bottles
HDPE, translucent.
  • Easy to fill due to wide opening
  • Cap absolutely tight closing
  • Suitable for use with food stuffs
Sturdy bottle with sealing eyelets on the screw cap and the bottle. Particularly suited for sending samples. Seals provide increased sample authenticity.