Sample cups, SeraNest™
Supplier: Simport Scientific
B720-13 B720-16
SIMPB720-13EA 617 EUR
SIMPB720-13 SIMPB720-16
Sample cups, SeraNest™
Sample Containers Sample Cups for Analysers
PS, clear.
  • Good chemical resistance
  • Good temperature resistance
These sample cups will hold perfectly on top of blood collection tubes. Just pour the blood sample into a SeraNest™ and its low shoulder will hold securely on the tube. No need to relabel. Two sizes available.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904