1,2,4,5-Tetrafluorobenzene ≥99%
Supplier: Thermo Fisher Scientific
|
Danger
|
A17815.06
A17815.14
A17815.06EA
74.5
EUR
A17815.06
A17815.14
1,2,4,5-Tetrafluorobenzene ≥99%
1,2,4,5-Tetrafluorobenzene
Purity: 99%
CAS No.: 327-54-8
Molecular Formula: C6H2F4
Formula Weight: 150.07
Flash Point: 16°(60°F)
Refractive Index: 1.4070
Melting Point: 3-5°
Boiling Point: 90°
Beilstein Reference: 1909052
UN No.: 2387
MDL No.: MFCD00000307
Formula:
C₆H₂F₄ Boiling Pt: 90 °C Melting Pt: 3…5 °C Density: 1.430 Flash Pt: 16 °C (60 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000307 CAS Number: 327-54-8 EINECS: 206-319-3 UN: 2387 ADR: 3,II |