2-Fluorobenzaldehyde 97%
Supplier: Thermo Fisher Scientific
Warning
119320250 119321000
ACRO119320250EA 53.2 EUR
ACRO119320250 ACRO119321000
2-Fluorobenzaldehyde 97%
2-Fluorobenzaldehyde
Formula: C₇H₅FO
MW: 124.11 g/mol
Boiling Pt: 172…174 °C
Melting Pt: –44.5 °C
Density: 1.17 g/cm³
Flash Pt: 55
Storage Temperature: Ambient
MDL Number: MFCD00003302
CAS Number: 446-52-6
EINECS: 207-171-2
UN: 1989
ADR: 3 III
Order Now
Specification Test Results
Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5205 to 1.5225 (20°C, 589 nm)
GC ≥96.0 %