2,3,4,5-Tetrafluorobenzoic acid
Supplier: Thermo Scientific
Warning
SB02010.1 SB02010.10 SB02010.50
MAYBSB02010.1EA 0 EUR
MAYBSB02010.1 MAYBSB02010.10 MAYBSB02010.50
2,3,4,5-Tetrafluorobenzoic acid
2,3,4,5-Tetrafluorobenzoic acid
Formula: C₇H₂F₄O₂
Boiling Pt: 135…137 °C (23 mmHg)
Melting Pt: 92…92.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00009613
CAS Number: 1201-31-6