1,3-Difluoro-4-iodobenzene ≥99%
Supplier: Thermo Fisher Scientific
|
Warning
|
A10798.09
A10798.18
A10798.09EA
34.9
EUR
A10798.09
A10798.18
1,3-Difluoro-4-iodobenzene ≥99%
1,3-Difluoro-4-iodobenzene
Purity: 99%
CAS No.: 2265-93-2
Molecular Formula: C6H3F2I
Formula Weight: 239.99
Flash Point: 67°(152°F)
Refractive Index: 1.5580
Boiling Point: 175-176°
Beilstein Reference: 2081078
MDL No.: MFCD00040151
Formula:
C₆H₃F₂I Boiling Pt: 175…176 °C Density: 2.006 Flash Pt: 67 °C (152 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040151 CAS Number: 2265-93-2 |