1-Fluoro-2-iodobenzene

Supplier: Thermo Scientific

Synonyms: 2-Fluoroiodobenzene

SB01067.1 SB01067.10 SB01067.50
MAYBSB01067.1EA 0 EUR
MAYBSB01067.1 MAYBSB01067.10 MAYBSB01067.50
1-Fluoro-2-iodobenzene
1-Fluoro-2-iodobenzene
Formula: C₆H₄FI
MW: 222 g/mol
Boiling Pt: 188
Melting Pt: –41.5 °C
Density: 1.903 g/cm³ (20 °C)
MDL Number: MFCD00001031
CAS Number: 348-52-7
EINECS: 206-477-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR