4-Fluoro-3-nitroaniline ≥98%
Supplier: Thermo Fisher Scientific
Danger
A15371.14 A15371.22 A15371.36
A15371.14EA 40.8 EUR
A15371.14 A15371.22 A15371.36
4-Fluoro-3-nitroaniline ≥98%
4-Fluoro-3-nitroaniline
Purity: 98% CAS No.: 364-76-1 Molecular Formula: C6H5FN2O2 Formula Weight: 156.12 Flash Point: 91°(195°F) Melting Point: 94-98° Beilstein Reference: 2210199 MDL No.: MFCD00007833
Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
Melting Pt: 94…98 °C
Density: 0.43 g/cm³ (20 °C)
Flash Pt: 91 °C (195 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007833
CAS Number: 364-76-1
EINECS: 206-665-5