3-Fluoroanisole ≥99%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
1-Fluoro-3-methoxybenzene
A10931.09
A10931.14
A10931.18
A10931.30
A10931.09EA
33.5
EUR
A10931.09
A10931.14
A10931.18
A10931.30
3-Fluoroanisole ≥99%
3-Fluoroanisole
Purity: 99%
CAS No.: 456-49-5
Molecular Formula: C7H7FO
Formula Weight: 126.13
Flash Point: 43°(109°F)
Refractive Index: 1.4890
Melting Point: -35°
Boiling Point: 158-159°
Beilstein Reference: 1858895
UN No.: 1993
MDL No.: MFCD00000335
Formula:
C₇H₇FO Boiling Pt: 158…159 °C Melting Pt: –35 °C Density: 1.104 Flash Pt: 43 °C (109 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000335 CAS Number: 456-49-5 EINECS: 207-267-4 UN: 1993 ADR: 3,III |