3,4-Difluorophenol ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
A12596.06
A12596.14
A12596.22
A12596.06EA
34.7
EUR
A12596.06
A12596.14
A12596.22
3,4-Difluorophenol ≥98%
3,4-Difluorophenol
Purity: 98%
CAS No.: 2713-33-9
Molecular Formula: C6H4F2O
Formula Weight: 130.09
Flash Point: 57°(134°F)
Odour: Phenol-like
Boiling Point: 84-86°/20mm
Beilstein Reference: 2081085
UN No.: 2921
MDL No.: MFCD00010315
Formula:
C₆H₄F₂O Boiling Pt: 84…86 °C (20 mmHg) Melting Pt: 34…38 °C Flash Pt: 57 °C (134 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00010315 CAS Number: 2713-33-9 EINECS: 424-300-1 UN: 2921 ADR: 8,II |