Iodobenzene 98%
Supplier: Thermo Fisher Scientific
Warning
122331000 122335000
ACRO122331000EA 114 EUR
ACRO122331000 ACRO122335000
Iodobenzene 98%
Iodobenzene
Formula: C₆H₅I
Boiling Pt: 188 °C (760 mmHg)
Melting Pt: –29 °C
Density: 1.82 g/cm³
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00001029
CAS Number: 591-50-4
EINECS: 209-719-6
Merck Index: 13,05051
Order Now
Specification Test Results
Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6180 to 1.6200 (20°C, 589 nm)
GC ≥97.5 %