2-Iodobenzaldehyde 98%
Supplier: Thermo Fisher Scientific
Warning
368170010 368170050
ACRO368170010EA 94.4 EUR
ACRO368170010 ACRO368170050
2-Iodobenzaldehyde 98%
2-Iodobenzaldehyde
Formula: C₇H₅IO
Boiling Pt: 129 °C (14 mmHg)
Melting Pt: 34…38 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039570
CAS Number: 26260-02-6
Order Now
Specification Test Results
Appearance White to light beige Powder or low melting crystalline mass
Infrared spectrum Conforms
Melting point 34 to 38 °C
GC ≥97.5 %