4-Ethoxyphenol 99%
Supplier: Thermo Fisher Scientific
Danger
409670250 409670050 409671000
ACRO409670250EA 61.3 EUR
ACRO409670250 ACRO409670050 ACRO409671000
4-Ethoxyphenol 99%
4-Ethoxyphenol
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 131 °C (9 mmHg)
Melting Pt: 64…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00002334
CAS Number: 622-62-8
EINECS: 210-748-1
Order Now
Specification Test Results
Appearance Beige Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 64 to 67 °C
GC ≥98.5 %