4-Chlororesorcinol, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
C70606-100G C70606-500G
SIALC70606-100GEA 64.6 EUR
SIALC70606-100G SIALC70606-500G
4-Chlororesorcinol, Sigma-Aldrich®
4-Chlororesorcinol
Formula: C₆H₅ClO₂
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 103…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00002273
CAS Number: 95-88-5
EINECS: 202-462-0