Guanine, Sigma-Aldrich®
Supplier: Merck
G6779-25G G6779-5G
SIALG6779-25GEA 747 EUR
SIALG6779-25G SIALG6779-5G
Guanine, Sigma-Aldrich®
Guanine
Formula: C₅H₅N₅O
Melting Pt: > 220 °C
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00071533
CAS Number: 73-40-5
EINECS: 200-799-8
Merck Index: 13,04580