D-(+)-Sucrose, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ300

S0389-1KG S0389-500G S0389-5KG
SIALS0389-1KGEA 210 EUR
SIALS0389-1KG SIALS0389-500G SIALS0389-5KG
D-(+)-Sucrose, Sigma-Aldrich®
D(+)-Sucrose
Formula: C₁₂H₂₂O₁₁
Melting Pt: 169…170 °C
Density: 1.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006626
CAS Number: 57-50-1
EINECS: 200-334-9
Merck Index: 13,08966
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR