2-Naphthalenesulphonic acid, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
249548-100G
249548-5G
SIAL249548-100GEA
122
EUR
SIAL249548-100G
SIAL249548-5G
2-Naphthalenesulphonic acid, Sigma-Aldrich®
2-Naphthalenesulphonic acid
Formula:
C₁₀H₈O₃S Boiling Pt: ∼350 °C (1013 hPa) Melting Pt: 124 °C Density: 1.441 g/cm³ (25 °C) |
MDL Number:
MFCD00149271 CAS Number: 120-18-3 EINECS: 204-375-3 Merck Index: 12,06464 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...