3,5-Dibromo-4-iodotoluene

Supplier: Thermo Scientific

TL00632.1 TL00632.10 TL00632.50
MAYBTL00632.1EA 0 EUR
MAYBTL00632.1 MAYBTL00632.10 MAYBTL00632.50
3,5-Dibromo-4-iodotoluene
3,5-Dibromo-4-iodotoluene
Formula: C₇H₅Br₂I
MDL Number: MFCD00052397
CAS Number: 175278-10-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR