1-Octene, Sigma-Aldrich®
Supplier: Merck
W429300-1KG W429300-5KG W429300-SAMPLE
SIALW429300-1KGEA 137 EUR
SIALW429300-1KG SIALW429300-5KG SIALW429300-SAMPLE
1-Octene, Sigma-Aldrich®
1-Octene
Formula: C₈H₁₆
Boiling Pt: 121 °C (1013 hPa)
Melting Pt: –102 °C
Density: 0.7152 g/cm³ (20 °C)
Flash Pt: 8 °C
Storage Temperature: Ambient
MDL Number: MFCD00009548
CAS Number: 111-66-0
EINECS: 203-893-7
UN: 3295
ADR: 3,II
Merck Index: 13,01770