1-Methyl-3-propylimidazolium iodide, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1-Methyl-3-n-propylimidazolium iodide, 1-Methyl-3-propyl-1H-imidazol-3-ium iodide
49637-50G 49637-5G
SIAL49637-50GEA 228 EUR
SIAL49637-50G SIAL49637-5G
1-Methyl-3-propylimidazolium iodide, Sigma-Aldrich®
1-Methyl-3-propylimidazolium iodide
Formula: C₇H₁₃IN₂
MW: 252.1 g/mol
MDL Number: MFCD08276385
CAS Number: 119171-18-5