Lithium chloride hydrate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Lithium chloride xhydrate, Hydrochloric acid lithium salt hydrate
M-Clarity™ quality level = MQ100
298328-100G 298328-25G
SIAL298328-100GEA 489 EUR
SIAL298328-100G SIAL298328-25G
Lithium chloride hydrate, Sigma-Aldrich®
Lithium chloride hydrate
Formula: ClLi·xH₂O
MW: 42.39 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00149764
CAS Number: 85144-11-2
EINECS: 231-212-3