1-Ethyl-3-methylimidazolium acetate, Sigma-Aldrich®

Supplier: Merck

51053-1KG-F 51053-100G-F
SIAL51053-1KG-FEA 1410 EUR
SIAL51053-1KG-F SIAL51053-100G-F
1-Ethyl-3-methylimidazolium acetate, Sigma-Aldrich®
Formula: C₈H₁₄N₂O₂
MDL Number: MFCD06798186
CAS Number: 143314-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR