Mycophenolic acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: (E)-6-(4-Hydroxy-6-methoxy-7-methyl-3-oxo-1,3-dihydroisobenzofuran-5-yl)-4-methylhex-4-enoic acid
M-Clarity™ quality level = MQ300
M5255-250MG M5255-50MG
SIALM5255-250MGEA 295 EUR
SIALM5255-250MG SIALM5255-50MG
Mycophenolic acid, Sigma-Aldrich®
Mycophenolic acid
Formula: C₁₇H₂₀O₆
MW: 320.34 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00036814
CAS Number: 24280-93-1
EINECS: 246-119-3