3,3',4,4'-Benzophenonetetracarboxylic dianhydride ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
A16529.22
A16529.36
A16529.A4
A16529.22EA
35.1
EUR
A16529.22
A16529.36
A16529.A4
3,3',4,4'-Benzophenonetetracarboxylic dianhydride ≥97%
3,3',4,4'-Benzophenonetetracarboxylic dianhydride
Purity: 97+%
CAS No.: 2421-28-5
Molecular Formula: C17H6O7
Formula Weight: 322.23
Flash Point: 324°(615°F)
Melting Point: 220-226°
Boiling Point: 320°/5mm
Beilstein Reference: 761777
MDL No.: MFCD00005923
Formula:
C₁₇H₆O₇ Boiling Pt: 320 °C (5 mmHg) Melting Pt: 220…226 °C Density: 1.570 Flash Pt: 324 °C (615 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005923 CAS Number: 2421-28-5 EINECS: 219-348-1 |