Geraniol, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: (E)-3,7-Dimethylocta-2,6-dien-1-ol

M-Clarity™ quality level = MQ300

W250708-1KG-K W250708-20KG-K W250708-9KG-K
SIALW250708-1KG-KEA 89.4 EUR
SIALW250708-1KG-K SIALW250708-20KG-K SIALW250708-9KG-K
Geraniol, Sigma-Aldrich®
Geraniol
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
MDL Number: MFCD00002917
CAS Number: 106-24-1
EINECS: 203-377-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR