Tryptamine hydrochloride, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
246557-25G 246557-5G
SIAL246557-25GEA 228 EUR
SIAL246557-25G SIAL246557-5G
Tryptamine hydrochloride, Sigma-Aldrich®
Tryptamine hydrochloride
Formula: C₁₀H₁₃ClN₂
MDL Number: MFCD00012682
CAS Number: 343-94-2
EINECS: 206-446-4