Ammonium acetate 7.5 M in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Synonyms: Acetic acid ammonium salt
M-Clarity™ quality level = MQ300
A2706-100ML A2706-1L A2706-500ML
SIALA2706-100MLEA 70.2 EUR
SIALA2706-100ML SIALA2706-1L SIALA2706-500ML
Ammonium acetate 7.5 M in aqueous solution, Sigma-Aldrich®
Ammonium acetate
Formula: H₃CCOONH₄
MW: 77.08 g/mol
Melting Pt: 114
Flash Pt: 136
Storage Temperature: Ambient
MDL Number: MFCD00013066
CAS Number: 631-61-8
EINECS: 211-162-9
Merck Index: 13,00495
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd160345483553318b94053de396335a9b18425f&vid=c6375b539008d6b43f50408d30b61261444faebc&prid=web&clid=SAPDAM