3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) ≥98%, Ultrapure
Supplier: VWR Chemicals
Warning
Synonyms: Thiazolyl blue tetrazolium bromide, MTT, 2-(4,5-Dimethylthiazol-2-yl)-3,5-diphenyl-2H-tetrazol-3-ium bromide
0793-1GEA 101 EUR
0793-1G 0793-5G
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) ≥98%, Ultrapure
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT)
MTT, a yellow tetrazole dye, can be reduced to a water-insoluble purple formazan compound by mitochondrial reductase enzymes. Since reduction only occurs in living cells, the quantitation of formazan can be equated to the number of viable cells in the population.
  • Sensitive colorimetric substitute for radioisotopes in cell proliferation and cytotoxicity studies
  • Applications include cell viability assays, dose response curves and cytotoxicity assays
Formula: C₁₈H₁₆N₅BrS
MW: 414.33 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00011964
CAS Number: 298-93-1
EINECS: 206-069-5
Order Now
Specification Test Results
Melting Range w/ Decomposition ≥187 °C
Purity ≥98%