2-Naphthalenethiol ≥98%
Supplier: Thermo Fisher Scientific
Danger
A12125.09 A12125.18
A12125.09EA 79.9 EUR
A12125.09 A12125.18
2-Naphthalenethiol ≥98%
2-Naphthalenethiol
Purity: 98+% CAS No.: 91-60-1 Molecular Formula: C10H8S Formula Weight: 160.24 Melting Point: 79-83° Boiling Point: 286° Merck Reference: 14,6379 MDL No.: MFCD00004086
Formula: C₁₀H₈S
Boiling Pt: 286 °C
Melting Pt: 79…83 °C
Density: 1.55
Storage Temperature: Ambient
MDL Number: MFCD00004086
CAS Number: 91-60-1
EINECS: 202-082-5
Merck Index: 14,6379