3-Aminophenylboronic acid pinacol ester, Sigma-Aldrich®

Supplier: Merck

Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 3-Aminobenzeneboronic acid pinacol ester

M-Clarity™ quality level = MQ100

574686-1G 574686-5G
SIAL574686-1GEA 29.3 EUR
SIAL574686-1G SIAL574686-5G
3-Aminophenylboronic acid pinacol ester, Sigma-Aldrich®
3-Aminophenylboronic acid pinacol ester
Formula: C₁₂H₁₈BNO₂
MDL Number: MFCD03453668
CAS Number: 210907-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR